Tisk…

2-Fluoro-3-nitrobenzylalcohol

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-Fluoro-3-nitrophenyl)methanol

PC501174-1G PC501174-250MG PC501174-5G
APOSPC501174-1GEA 1210 CZK
APOSPC501174-1G APOSPC501174-250MG APOSPC501174-5G
2-Fluoro-3-nitrobenzylalcohol
2-Fluoro-3-nitrobenzylalcohol
Vzorec: C₇H₆FNO₃
MW: 171,1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835208
CAS číslo: 946126-95-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR