Tisk…

5-Chloro-4-fluoro-2-nitrophenol

Dodavatel: Apollo Scientific

PC501173-5G
APOSPC501173-5GEA 3190 CZK
APOSPC501173-5G
5-Chloro-4-fluoro-2-nitrophenol
5-Chloro-4-fluoro-2-nitrophenol
Vzorec: C6H3ClFNO3
MW: 191,5400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18072578
CAS číslo: 65001-79-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR