Tisk…
Lithium trifluoroacetate 97%
Dodavatel: Apollo Scientific
Varování
PC5011-100G PC5011-10G PC5011-50G
APOSPC5011-100GEA 6290 CZK
APOSPC5011-100G APOSPC5011-10G APOSPC5011-50G
Lithium trifluoroacetate 97%
Lithium trifluoroacetate
Useful for the preparation of trifluoroketones; treatment with BuLi gives 61% yield of 1,1,1-trifluoro-2-hexanone;also reacts with Grignard reagents to give ketones.
Vzorec: LiCO₂CF₃
MW: 119.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013216
CAS číslo: 2923-17-3
EINECS: 220-878-0