3-(Pentafluorothio)benzyl bromide 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC50105-1G
PC50105-5G
APOSPC50105-1GEA
14680
CZK
APOSPC50105-1G
APOSPC50105-5G
3-(Pentafluorothio)benzyl bromide 97%
3-(Pentafluorothio)benzyl bromide
Vzorec:
C₇H₆BrF₅S MW: 297,08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16652411 CAS číslo: 1056893-24-3 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...