Tisk…

2-Chloro-5-fluoro-3-nitropyridine

Dodavatel: Apollo Scientific

PC50086-25G PC50086-5G
APOSPC50086-25GEA 6950 CZK
APOSPC50086-25G APOSPC50086-5G
2-Chloro-5-fluoro-3-nitropyridine
2-Chloro-5-fluoro-3-nitropyridine
Vzorec: C₅H₂ClFN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06659490
CAS číslo: 136888-21-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR