3-Bromo-2,6-difluoropyridine
Dodavatel: Apollo Scientific
|
Varování
|
PC500796-1G
PC500796-5G
APOSPC500796-1GEA
2330
CZK
APOSPC500796-1G
APOSPC500796-5G
3-Bromo-2,6-difluoropyridine
3-Bromo-2,6-difluoropyridine
Vzorec:
C₅H₂BrF₂N MW: 193.98 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11044274 CAS číslo: 80392-79-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...