Tisk…

(2-Fluoro-4-methoxyphenyl)acetonitrile 97%

Dodavatel: Apollo Scientific

PC500442-1G PC500442-5G
APOSPC500442-1GEA 3310 CZK
APOSPC500442-1G APOSPC500442-5G
(2-Fluoro-4-methoxyphenyl)acetonitrile 97%
(2-Fluoro-4-methoxyphenyl)acetonitrile
Vzorec: C₉H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD09038467
CAS číslo: 749934-29-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR