6-Chloro-5-(trifluoromethyl)pyridine-3-carboxylic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC50039-100MG
PC50039-1G
PC50039-250MG
APOSPC50039-100MGEA
3940
CZK
APOSPC50039-100MG
APOSPC50039-1G
APOSPC50039-250MG
6-Chloro-5-(trifluoromethyl)pyridine-3-carboxylic acid 98%
6-Chloro-5-(trifluoromethyl)pyridine-3-carboxylic acid
Vzorec:
C₇H₃ClF₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD11848376 CAS číslo: 1110782-41-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...