3-Fluoro-4-nitrobenzenesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3-Fluoro-4-nitrobenzene-1-sulphonyl chloride
PC500352-1G
PC500352-25G
PC500352-5G
APOSPC500352-1GEA
1160
CZK
APOSPC500352-1G
APOSPC500352-25G
APOSPC500352-5G
3-Fluoro-4-nitrobenzenesulphonyl chloride
3-Fluoro-4-nitrobenzenesulphonyl chloride
Vzorec:
C₆H₃ClFNO₄S MW: 239,61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03412240 CAS číslo: 86156-93-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...