Tisk…

5-Bromo-2-chloro-4-fluorophenol

Dodavatel: Apollo Scientific
Varování

PC500254-1G PC500254-250MG PC500254-5G
APOSPC500254-1GEA 744 CZK
APOSPC500254-1G APOSPC500254-250MG APOSPC500254-5G
5-Bromo-2-chloro-4-fluorophenol
5-Bromo-2-chloro-4-fluorophenol
Vzorec: C₆H₃BrClFO
MW: 225,4428 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08436979
CAS číslo: 148254-32-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR