4-Bromo-5-fluoro-2-methoxyaniline
Dodavatel: Apollo Scientific
|
Varování
|
PC500184-25G
PC500184-5G
APOSPC500184-25GEA
0
CZK
APOSPC500184-25G
APOSPC500184-5G
4-Bromo-5-fluoro-2-methoxyaniline
4-Bromo-5-fluoro-2-methoxyaniline
Vzorec:
C₇H₇BrFNO Storage Temperature: Ambient |
MDL Number:
MFCD21603958 CAS číslo: 330794-03-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...