Tisk…

3-Chloro-4-(trifluoromethyl)phenol 98%

Dodavatel: Apollo Scientific
Varování

PC500172-1G PC500172-250MG PC500172-5G
APOSPC500172-1GEA 3540 CZK
APOSPC500172-1G APOSPC500172-250MG APOSPC500172-5G
3-Chloro-4-(trifluoromethyl)phenol 98%
3-Chloro-4-(trifluoromethyl)phenol
Vzorec: C₇H₄ClF₃O
Storage Temperature: Refrigerator
MDL Number: MFCD11849933
CAS číslo: 37900-81-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR