4-Fluoro-3-(trifluoromethoxy)phenol
Dodavatel: Apollo Scientific
|
Varování
|
PC500166-1G
PC500166-5G
PC500166-25G
APOSPC500166-1GEA
4370
CZK
APOSPC500166-1G
APOSPC500166-5G
APOSPC500166-25G
4-Fluoro-3-(trifluoromethoxy)phenol
4-Fluoro-3-(trifluoromethoxy)phenol
Vzorec:
C₇H₄F₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD06660257 CAS číslo: 886501-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...