(3-(Trifluoromethyl)phenyl)methanesulphonyl chloride
Dodavatel: Apollo Scientific
Synonyms:
3-(Trifluoromethyl)benzylsulphonyl chloride
PC50013-1G
PC50013-250MG
PC50013-500MG
PC50013-5G
APOSPC50013-1GEA
1370
CZK
APOSPC50013-1G
APOSPC50013-250MG
APOSPC50013-500MG
APOSPC50013-5G
(3-(Trifluoromethyl)phenyl)methanesulphonyl chloride
(3-(Trifluoromethyl)phenyl)methanesulphonyl chloride
Vzorec:
C₈H₆ClF₃O₂S Bod tání: 59…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631926 CAS číslo: 127162-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...