Tisk…

1-Bromo-3-(difluoromethyl)benzene 98%

Dodavatel: Apollo Scientific
Varování

PC50007-100G PC50007-25G PC50007-5G
APOSPC50007-100GEA 12830 CZK
APOSPC50007-100G APOSPC50007-25G APOSPC50007-5G
1-Bromo-3-(difluoromethyl)benzene 98%
1-Bromo-3-(difluoromethyl)benzene
Vzorec: C₇H₅BrF₂
Storage Temperature: Ambient
MDL Number: MFCD07368984
CAS číslo: 29848-59-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR