3-bromo-2,6-difluoroaniline 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC500008-1G
PC500008-250MG
PC500008-5G
APOSPC500008-1GEA
705
CZK
APOSPC500008-1G
APOSPC500008-250MG
APOSPC500008-5G
3-bromo-2,6-difluoroaniline 95%
3-Bromo-2,6-difluoroaniline
Vzorec:
C₆H₄BrF₂N MW: 208.006 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22627833 CAS číslo: 1262198-07-1 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...