2',3'-Difluoroacetophenone 97%
Dodavatel: Apollo Scientific
Synonyms:
1-(2,3-Difluorophenyl)ethan-1-one, 1-(2,3-Difluorophenyl)ethanone, 2,3-Difluoroacetophenone
PC49704-100G
PC49704-25G
PC49704-5G
APOSPC49704-100GEA
7180
CZK
APOSPC49704-100G
APOSPC49704-25G
APOSPC49704-5G
2',3'-Difluoroacetophenone 97%
2',3'-Difluoroacetophenone
Vzorec:
C₈H₆F₂O MW: 156.13 Storage Temperature: Ambient |
MDL Number:
MFCD00061274 CAS číslo: 18355-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...