Tisk…

5-Bromo-2-chloro-3-(difluoromethyl)pyridine 97%

Dodavatel: Apollo Scientific
Varování

PC49672-1G PC49672-5G
APOSPC49672-1GEA 7440 CZK
APOSPC49672-1G APOSPC49672-5G
5-Bromo-2-chloro-3-(difluoromethyl)pyridine 97%
5-Bromo-2-chloro-3-(difluoromethyl)pyridine
Vzorec: C₆H₃BrClF₂N
Storage Temperature: Ambient
MDL Number: MFCD22199307
CAS číslo: 1332392-76-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR