2-Fluoropropenoic acid 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC4962-5G
PC4962-1G
APOSPC4962-5GEA
3150
CZK
APOSPC4962-5G
APOSPC4962-1G
2-Fluoropropenoic acid 97%
2-Fluoropropenoic acid
Vzorec:
C₃H₃FO₂ Bod tání: 51…52 °C Storage Temperature: Freezer |
MDL Number:
MFCD03424474 CAS číslo: 430-99-9 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...