5-Bromo-7-(trifluoromethyl)quinoxaline
Dodavatel: Apollo Scientific
|
Varování
|
PC49608-1G
PC49608-250MG
PC49608-500MG
PC49608-5G
APOSPC49608-1GEA
14780
CZK
APOSPC49608-1G
APOSPC49608-250MG
APOSPC49608-500MG
APOSPC49608-5G
5-Bromo-7-(trifluoromethyl)quinoxaline
5-Bromo-7-(trifluoromethyl)quinoxaline
Vzorec:
C₉H₄BrF₃N₂ Storage Temperature: Ambient |
MDL Number:
MFCD18089243 CAS číslo: 1287218-49-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...