Tisk…

Lanthanum(III) fluoride, anhydrous 99,9%

Dodavatel: Apollo Scientific
Varování

Synonyms: Lanthanum trifluoride

PC4960-100G PC4960-500G
APOSPC4960-100GEA 4170 CZK
APOSPC4960-100G APOSPC4960-500G
Lanthanum(III) fluoride, anhydrous 99,9%
Lanthanum(III) fluoride
Vzorec: F₃La
MW: 195,9 g/mol
Bod varu: 2230 °C
Bod tání: 1493 °C
Hustota: 5,936 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011073
CAS číslo: 13709-38-1
EINECS: 237-252-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR