Methyl-4-amino-3-fluoro-5-iodobenzoate 95+%
Dodavatel: Apollo Scientific
PC49533-1G
PC49533-250MG
PC49533-5G
APOSPC49533-1GEA
8100
CZK
APOSPC49533-1G
APOSPC49533-250MG
APOSPC49533-5G
Methyl-4-amino-3-fluoro-5-iodobenzoate 95+%
Methyl-4-amino-3-fluoro-5-iodobenzoate
Vzorec:
C₈H₇FINO₂ MW: 295,05 g/mol Bod tání: 115…117 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD13176511 CAS číslo: 1123172-01-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...