Tisk…

5-Bromo-2-methox-3-(trifluoromethyl)ypyridine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-Bromo-2-methoxy-3-(trifluoromethyl)pyridine

PC49527-1G PC49527-5G PC49527-10G
APOSPC49527-1GEA 622 CZK
APOSPC49527-1G APOSPC49527-5G APOSPC49527-10G
5-Bromo-2-methox-3-(trifluoromethyl)ypyridine
5-Bromo-2-methox-3-(trifluoromethyl)ypyridine
Vzorec: C₇H₅BrF₃NO
MW: 256,0199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185815
CAS číslo: 1214377-42-0
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR