Tisk…

4-(Trifluoroacetyl)piperidine hydrochloride

Dodavatel: Apollo Scientific
Varování

PC49504-1G PC49504-250MG
APOSPC49504-1GEA 23970 CZK
APOSPC49504-1G APOSPC49504-250MG
4-(Trifluoroacetyl)piperidine hydrochloride
4-(Trifluoroacetyl)piperidine hydrochloride
Vzorec: C₇H₁₁ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD22041637
CAS číslo: 1372103-92-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR