Tisk…

5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazole carbohydrazide 97%

Dodavatel: Apollo Scientific

PC4950-1G PC4950-500MG
APOSPC4950-1GEA 30060 CZK
APOSPC4950-1G APOSPC4950-500MG
5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazole carbohydrazide 97%
5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazole carbohydrazide
Vzorec: C₁₁H₁₁ClF₃N₅O₂
Bod tání: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00202621
CAS číslo: 251310-32-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR