2-{1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-indol-3-yl}acetonitrile 97%
Dodavatel: Apollo Scientific
PC4943-1G
PC4943-500MG
APOSPC4943-1GEA
22750
CZK
APOSPC4943-1G
APOSPC4943-500MG
2-{1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-indol-3-yl}acetonitrile 97%
2-{1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-indol-3-yl}acetonitrile
Vzorec:
C₁₆H₉ClF₃N₃ MW: 335,72 g/mol Bod tání: 104…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00664465 CAS číslo: 303152-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...