Tisk…

2-Iodo-4-(trifluoromethyl)benzoic acid 97%

Dodavatel: Apollo Scientific
Varování

PC49379-1G PC49379-5G PC49379-25G
APOSPC49379-1GEA 787 CZK
APOSPC49379-1G APOSPC49379-5G APOSPC49379-25G
2-Iodo-4-(trifluoromethyl)benzoic acid 97%
2-Iodo-4-(trifluoromethyl)benzoic acid
Vzorec: C₈H₄F₃IO₂
MW: 316,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06408817
CAS číslo: 54507-44-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR