1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-N-(2-hydroxyethyl)piperdine-4-carboxamide 97%
Dodavatel: Apollo Scientific
PC4934-1G
PC4934-500MG
PC4934-5G
APOSPC4934-1GEA
15180
CZK
APOSPC4934-1G
APOSPC4934-500MG
APOSPC4934-5G
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-N-(2-hydroxyethyl)piperdine-4-carboxamide 97%
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-N-(2-hydroxyethyl)piperdine-4-carboxamide
Vzorec:
C₁₄H₁₇ClF₃N₃O₂ MW: 351,76 g/mol Bod tání: 174…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD00172258 CAS číslo: 303150-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...