(2-Fluorobenzyl)hydrazine 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC49334-10G
PC49334-25G
PC49334-5G
APOSPC49334-10GEA
4070
CZK
APOSPC49334-10G
APOSPC49334-25G
APOSPC49334-5G
(2-Fluorobenzyl)hydrazine 95%
(2-Fluorobenzyl)hydrazine
Vzorec:
C₇H₉FN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD07785903 CAS číslo: 51859-98-4 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...