Tisk…

4-Chloro-3-(trifluoromethoxy)benzyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC49329-1G PC49329-5G PC49329-25G
APOSPC49329-1GEA 1780 CZK
APOSPC49329-1G APOSPC49329-5G APOSPC49329-25G
4-Chloro-3-(trifluoromethoxy)benzyl bromide 97%
4-Chloro-3-(trifluoromethoxy)benzyl bromide
Vzorec: C₈H₅BrClF₃O
Storage Temperature: Ambient
MDL Number: MFCD06660249
CAS číslo: 886500-93-2
UN: 3265
ADR: 8,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR