Tisk…

Methyl-2-(trifluoromethyl)isonicotinate 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 2-(trifluoromethyl)isonicotinate

PC49313-250MG PC49313-1G
APOSPC49313-250MGEA 1490 CZK
APOSPC49313-250MG APOSPC49313-1G
Methyl-2-(trifluoromethyl)isonicotinate 95%
Methyl-2-(trifluoromethyl)isonicotinate
Vzorec: C₈H₆F₃NO₂
MW: 205,136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698093
CAS číslo: 588702-68-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR