Tisk…

Methyl-2-fluoro-3-nitrobenzoate 99%

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 2-fluoro-3-nitrobenzoate

PC49297-1G PC49297-25G PC49297-5G
APOSPC49297-1GEA 622 CZK
APOSPC49297-1G APOSPC49297-25G APOSPC49297-5G
Methyl-2-fluoro-3-nitrobenzoate 99%
Methyl-2-fluoro-3-nitrobenzoate
Vzorec: C₈H₆FNO₄
MW: 199,1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566813
CAS číslo: 946126-94-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR