4-Fluoro-3-(trifluoromethoxy)benzyl bromide 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC49274-1G
PC49274-5G
PC49274-10G
APOSPC49274-1GEA
6420
CZK
APOSPC49274-1G
APOSPC49274-5G
APOSPC49274-10G
4-Fluoro-3-(trifluoromethoxy)benzyl bromide 98%
4-Fluoro-3-(trifluoromethoxy)benzyl bromide
Vzorec:
C₈H₅BrF₄O MW: 273,02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06660256 CAS číslo: 86256-50-0 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...