2,3-Difluoro-4-iodobenzaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
PC49271-1G
PC49271-5G
APOSPC49271-1GEA
3270
CZK
APOSPC49271-1G
APOSPC49271-5G
2,3-Difluoro-4-iodobenzaldehyde
2,3-Difluoro-4-iodobenzaldehyde
Vzorec:
C₇H₃F₂IO MW: 268,0000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09835190 CAS číslo: 885590-99-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...