Tisk…

3-(Difluoromethyl)benzaldehyde 97%

Dodavatel: Apollo Scientific
Varování

PC49198-1G PC49198-5G
APOSPC49198-1GEA 4570 CZK
APOSPC49198-1G APOSPC49198-5G
3-(Difluoromethyl)benzaldehyde 97%
3-(Difluoromethyl)benzaldehyde
Vzorec: C₈H₆F₂O
Storage Temperature: Ambient
MDL Number: MFCD18791599
CAS číslo: 945004-44-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR