Tisk…

4-(Trifluoromethyl)-3-pyridineboronic acid pinacol ester

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(Trifluoromethyl)pyridine-3-boronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyridine

PC49188-100MG PC49188-1G PC49188-250MG
APOSPC49188-100MGEA 622 CZK
APOSPC49188-100MG APOSPC49188-1G APOSPC49188-250MG
4-(Trifluoromethyl)-3-pyridineboronic acid pinacol ester
4-(Trifluoromethyl)-3-pyridineboronic acid pinacol ester
Vzorec: C₁₂H₁₅BF₃NO₂
MW: 273,05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12198140
CAS číslo: 1310405-06-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR