Tisk…

4-Fluoro-3-nitrophenol 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC49179-1G PC49179-25G PC49179-5G
APOSPC49179-1GEA 622 CZK
APOSPC49179-1G APOSPC49179-25G APOSPC49179-5G
4-Fluoro-3-nitrophenol 97%
4-Fluoro-3-nitrophenol
Vzorec: C₆H₄FNO₃
MW: 157,0993 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04038714
CAS číslo: 2105-96-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR