[1-(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)-2-sulphanyl-1H-imidazol-5-yl]methanol 97%
Dodavatel: Apollo Scientific
Synonyms:
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylamino]ethyl-5-(hydroxymethyl)-1H-imidazole-2-thiol
PC4917-1G
PC4917-500MG
APOSPC4917-1GEA
18980
CZK
APOSPC4917-1G
APOSPC4917-500MG
[1-(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)-2-sulphanyl-1H-imidazol-5-yl]methanol 97%
[1-(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)-2-sulphanyl-1H-imidazol-5-yl]methanol
Vzorec:
C₁₂H₁₂ClF₃N₄OS Bod tání: 219…221 °C Storage Temperature: Ambient |
MDL Number:
MFCD00215163 CAS číslo: 338794-80-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...