Potassium trifluoro(prop-1-en-2-yl)borate 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Potassium Isopropenyltrifluoroborate
PC49166-5G
PC49166-25G
PC49166-100G
APOSPC49166-5GEA
622
CZK
APOSPC49166-5G
APOSPC49166-25G
APOSPC49166-100G
Potassium trifluoro(prop-1-en-2-yl)borate 95%
Potassium trifluoro(prop-1-en-2-yl)borate
Vzorec:
C3H5BF3K MW: 147,9763 g/mol Bod tání: 274…280 °C Storage Temperature: Ambient |
MDL Number:
MFCD11052654 CAS číslo: 395083-14-4 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...