2-Bromo-3-fluoro-6-methylpyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Bromo-3-fluoro-6-picoline
PC49163-1G
PC49163-5G
APOSPC49163-1GEA
622
CZK
APOSPC49163-1G
APOSPC49163-5G
2-Bromo-3-fluoro-6-methylpyridine
2-Bromo-3-fluoro-6-methylpyridine
Vzorec:
C₆H₅BrFN MW: 190,01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03095061 CAS číslo: 374633-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...