Tisk…

3-Bromo-5-fluoropicolinonitrile

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Bromo-5-fluoro-2-cyanopyridine

PC49162-1G PC49162-250MG
APOSPC49162-1GEA 7250 CZK
APOSPC49162-1G APOSPC49162-250MG
3-Bromo-5-fluoropicolinonitrile
3-Bromo-5-fluoropicolinonitrile
Vzorec: C₆H₂BrFN₂
MW: 201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16988825
CAS číslo: 950670-18-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR