Ytterbium(III) trifluoromethanesulphonate hydrate 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Trifluoromethanesulphonic acid ytterbium salt hydrate, Ytterbium(III) trifluoromethanesulphonate hydrate, Ytterbium(III) triflate hydrate, Ytterbium triflate hydrate
PC49155-10G
PC49155-25G
APOSPC49155-10GEA
2530
CZK
APOSPC49155-10G
APOSPC49155-25G
Ytterbium(III) trifluoromethanesulphonate hydrate 98%
Ytterbium(III) trifluoromethanesulphonate hydrate
Vzorec:
C₃F₉O₉S₃Yb·xH₂O Bod tání: 118…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD03703499 CAS číslo: 252976-51-5 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...