3-(Difluoromethyl)aniline 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC49148-1G
PC49148-250MG
PC49148-5G
APOSPC49148-1GEA
15180
CZK
APOSPC49148-1G
APOSPC49148-250MG
APOSPC49148-5G
3-(Difluoromethyl)aniline 95%
3-(Difluoromethyl)aniline
Vzorec:
C₇H₇F₂N MW: 143,137 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18384830 CAS číslo: 368-99-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...