2-Bromo-4-(difluoromethyl)pyridine
Dodavatel: Apollo Scientific
PC49139-1G
PC49139-500MG
APOSPC49139-1GEA
7540
CZK
APOSPC49139-1G
APOSPC49139-500MG
2-Bromo-4-(difluoromethyl)pyridine
2-Bromo-4-(difluoromethyl)pyridine
Vzorec:
C₆H₄BrF₂N Hustota: 1,6573 g/cm³ (25 °C) Bod vzplanutí: 103 °C Storage Temperature: Ambient |
MDL Number:
MFCD14525493 CAS číslo: 1204295-87-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...