Tisk…

2-Fluoro-4-methylbenzenesulphonamide

Dodavatel: Apollo Scientific
Varování

PC49099-1G PC49099-5G
APOSPC49099-1GEA 3870 CZK
APOSPC49099-1G APOSPC49099-5G
2-Fluoro-4-methylbenzenesulphonamide
2-Fluoro-4-methylbenzenesulphonamide
Vzorec: C₇H₈FNO₂S
Storage Temperature: Ambient
MDL Number: MFCD09800666
CAS číslo: 199590-69-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR