5-Bromo-3-fluoro-2-methoxypyridine 97%
Dodavatel: Apollo Scientific
PC49087-25G
PC49087-5G
APOSPC49087-25GEA
5530
CZK
APOSPC49087-25G
APOSPC49087-5G
5-Bromo-3-fluoro-2-methoxypyridine 97%
5-Bromo-3-fluoro-2-methoxypyridine
Vzorec:
C₆H₅BrFNO Storage Temperature: Ambient |
MDL Number:
MFCD04112553 CAS číslo: 124432-70-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...