Ethyl-2-[(2-fluorobenzoyl)amino]acetate 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC4897-5G
APOSPC4897-5GEA
9100
CZK
APOSPC4897-5G
Ethyl-2-[(2-fluorobenzoyl)amino]acetate 97%
Ethyl-2-[(2-fluorobenzoyl)amino]acetate
Vzorec:
C₁₁H₁₂FNO₃ MW: 225,22 g/mol Bod tání: 40…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00793744 CAS číslo: 304657-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...