α-Bromo-2-(difluoromethoxy)toluene, Technical Grade
Dodavatel: Apollo Scientific
Synonyms:
o-(Difluoromethoxy)benzyl bromide, 1-(Bromomethyl)-2-(difluoromethoxy)benzene, 2-(Difluoromethoxy)benzyl bromide
PC48840-100G
PC48840-25G
PC48840-5G
APOSPC48840-100GEA
9100
CZK
APOSPC48840-100G
APOSPC48840-25G
APOSPC48840-5G
α-Bromo-2-(difluoromethoxy)toluene, Technical Grade
α-Bromo-2-(difluoromethoxy)toluene
Vzorec:
C₈H₇BrF₂O MW: 237.04 Bod varu: 227 Bod tání: 13 Hustota: 1.555 Storage Temperature: Ambient |
MDL Number:
MFCD00190103 CAS číslo: 85684-64-6 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...