Tisk…

Ethyl-2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}-3-(dimethylamino)acrylate

Dodavatel: Apollo Scientific

PC4884-1G PC4884-500MG PC4884-5G
APOSPC4884-1GEA 15180 CZK
APOSPC4884-1G APOSPC4884-500MG APOSPC4884-5G
Ethyl-2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}-3-(dimethylamino)acrylate
Ethyl-2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}-3-(dimethylamino)acrylate
Vzorec: C₁₃H₁₄ClF₃N₂O₂S
MW: 354,78 g/mol
Bod tání: 102…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00664271
CAS číslo: 338422-75-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR