Tisk…

Ethyl [3-chloro-5-(trifluoromethyl)-pyridin-2-yl]thio}acetate

Dodavatel: Apollo Scientific

PC4883-1G PC4883-5G
APOSPC4883-1GEA 1820 CZK
APOSPC4883-1G APOSPC4883-5G
Ethyl [3-chloro-5-(trifluoromethyl)-pyridin-2-yl]thio}acetate
Ethyl [3-chloro-5-(trifluoromethyl)-pyridin-2-yl]thio}acetate
Vzorec: C₁₀H₉ClF₃NO₂S
MW: 299,7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00664270
CAS číslo: 338422-70-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR